DL-Carnitin Hydrochlorid ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride
A16589.14 A16589.22
A16589.14EA 28.8 EUR
A16589.14 A16589.22
DL-Carnitin Hydrochlorid ≥98%
DL-Carnitin Hydrochlorid
Purity: 98+% CAS No.: 461-05-2 Molecular Formula: C7H15NO3.HCl Formula Weight: 197.66 Melting Point: 196-200° Beilstein Reference: 4163212 Merck Reference: 14,1849 MDL No.: MFCD00011904
Formel: C₇H₁₆ClNO₃
Molecular Weight: 197,66 g/mol
Schmelzpunkt: 196…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011904
CAS-Nummer: 461-05-2
EINECS: 207-309-1
Merck Index: 14,1849