L(-)-Carnitin Hydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Levocarnitine hydrochloride, L-Carnitine hydrochloride, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride, LC-80, (R)-3-Hydroxy-4-(trimethylammonio)butyrate hydrochloride, L-Carnitine chloride, (R)-Carnitine hydrochloride, Levocarnitine chloride

230280050 230280250 230281000
ACRO230280050EA 43 EUR
ACRO230280050 ACRO230280250 ACRO230281000
L(-)-Carnitin Hydrochlorid 98%
L(-)-Carnitin Hydrochlorid
Formel: C₇H₁₆ClNO₃
Molecular Weight: 197,66 g/mol
Schmelzpunkt: 142 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00066100
CAS-Nummer: 6645-46-1
EINECS: 229-663-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Sulfated ash ≤0.5 %
Specific optical rotation -21° to -24° (20°C, 589 nm) (c=1, H2O)
Titration with NaOH 97.5 to 102.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR