(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: CTH-(S)-Xylyl-P-Phos, (S)-4,4'-Bis(bis(3,5-dimethylphenyl)phosphino)-2,2',6,6'-tetramethoxy-3,3'-bipyridine

M-Clarity™ Qualitätsstufe = MQ100

675946-100MG 675946-500MG
SIAL675946-100MGEA 105 EUR
SIAL675946-100MG SIAL675946-500MG
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridin, Sigma-Aldrich®
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridin
Formel: C₄₆H₅₀N₂O₄P₂
Molecular Weight: 756,85 g/mol
MDL: MFCD04974235
CAS-Nummer: 443347-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR