Indometacin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

I8280-100G I8280-25G I8280-5G
SIALI8280-100GEA 1350 EUR
SIALI8280-100G SIALI8280-25G SIALI8280-5G
Indometacin, Sigma-Aldrich®
Indomethacin
Formel: C₁₉H₁₆ClNO₄
Molecular Weight: 357.80 g/mol
MDL: MFCD00057095
CAS-Nummer: 53-86-1
EINECS: 200-186-5
UN: 2811
ADR: 6.1 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR