3-(2-Benzothiazolyl)-7-(diethylamino)cumarin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(Benzo[d]thiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one, Coumarin 6
M-Clarity™ Qualitätsstufe = MQ100
442631-1G
442631-5G
SIAL442631-1GEA
92.9
EUR
SIAL442631-1G
SIAL442631-5G
3-(2-Benzothiazolyl)-7-(diethylamino)cumarin, Sigma-Aldrich®
3-(2-Benzothiazolyl)-7-(diethylamino)cumarin
Formel:
C₂₀H₁₈N₂O₂S Molecular Weight: 350,4341 g/mol |
MDL:
MFCD00041869 CAS-Nummer: 38215-36-0 EINECS: 253-830-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...