Iminodibenzyl 97%
Lieferant: Thermo Scientific
Synonyme:
10,11-Dihydro-5H-dibenzo[b,f]azepine
A19204.14
A19204.22
A19204.36
A19204.14EA
33.7
EUR
A19204.14
A19204.22
A19204.36
Iminodibenzyl 97%
Iminodibenzyl
Purity: 97%
CAS No.: 494-19-9
Molecular Formula: C14H13N
Formula Weight: 195.27
Odour: Odourless
Melting Point: 105-108°
Beilstein Reference: 152732
MDL No.: MFCD00005070
Formel:
C₁₄H₁₃N Molecular Weight: 195,27 g/mol Schmelzpunkt: 105…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005070 CAS-Nummer: 494-19-9 EINECS: 207-787-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...