Diphenylborsäure(2-aminoethyl)ester (2-APB), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine

D9754-10G D9754-25G D9754-1G D9754-5G
SIALD9754-10GEA 164 EUR
SIALD9754-10G SIALD9754-25G SIALD9754-1G SIALD9754-5G
Diphenylborsäure(2-aminoethyl)ester (2-APB), Sigma-Aldrich®
Diphenylborsäure(2-aminoethyl)ester (2-APB)
Formel: C₁₄H₁₆BNO
Molecular Weight: 225.1 g/mol
MDL: MFCD00014823
CAS-Nummer: 524-95-8
EINECS: 208-366-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR