Luminol (3-Aminophthalhydrazid)
Lieferant: BIOMOL RESEARCH LABORATORIES
Synonyme: NSC 5064
54570-2 54570-3
BMOL54570-2EA 0 EUR
BMOL54570-2 BMOL54570-3
Luminol (3-Aminophthalhydrazid)
Luminol (3-Aminophthalhydrazid)
Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
MDL: MFCD00006890
CAS-Nummer: 521-31-3
EINECS: 208-309-4