Phthalazin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Benzo[d]pyridazine

A12270.06 A12270.14
A12270.06EA 48.9 EUR
A12270.06 A12270.14
Phthalazin ≥98%
Phthalazine

Purity: 98% CAS No.: 253-52-1 Molecular Formula: C8H6N2 Formula Weight: 130.15 Melting Point: 89-92° Boiling Point: 317° Merck Reference: 14,7370 MDL No.: MFCD00006908

Formel: C₈H₆N₂
Molecular Weight: 130.15 g/mol
Siedepunkt: 317 °C
Schmelzpunkt: 89…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006908
CAS-Nummer: 253-52-1
EINECS: 205-963-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR