Phthalazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzo[d]pyridazine

M-Clarity™ Qualitätsstufe = MQ100

P38706-10G P38706-1G
SIALP38706-10GEA 125 EUR
SIALP38706-10G SIALP38706-1G
Phthalazin, Sigma-Aldrich®
Phthalazine
Formel: C₈H₆N₂
Molecular Weight: 130.15 g/mol
MDL: MFCD00006908
CAS-Nummer: 253-52-1
EINECS: 205-963-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR