L-Methionin-[Methyl-13C,Methyl-D3] (99% ¹³C , 99% D), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
L-Methionine-[methyl-¹³C,methyl-²H₃]
M-Clarity™ Qualitätsstufe = MQ200
299154-1G
299154-250MG
SIAL299154-1GEA
1140
EUR
SIAL299154-1G
SIAL299154-250MG
L-Methionin-[Methyl-13C,Methyl-D3] (99% ¹³C , 99% D), Sigma-Aldrich®
L-Methionin-[Methyl-13C,Methyl-D3]
Formel:
C₅H₈D₃NO₂S Lagertemperatur: Raumtemperatur |
CAS-Nummer:
73488-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...