(4-Aminophenyl)essigsäure

Lieferant: CHEMPUR

Synonyme: 4-Aminophenylacetic acid

BTB07663.250 BTB07663.1
CHMPBTB07663.250EA 0 EUR
CHMPBTB07663.250 MAYBBTB07663.1
(4-Aminophenyl)essigsäure
(4-Aminophenyl)essigsäure
Formel: (C₆H₄(NH₂))CH₂COOH
Molecular Weight: 151,16 g/mol
Siedepunkt: 322 °C (1013 hPa)
Schmelzpunkt: 199…202 °C
MDL: MFCD00007916
CAS-Nummer: 1197-55-3
EINECS: 214-828-7
Merck Index: 13,00462

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR