(1,1,2,2-Tetrafluorethoxy)benzol

Lieferant: CHEMPUR
Gefahr

RJC02403.250 RJC02403.1
CHMPRJC02403.250EA 0 EUR
CHMPRJC02403.250 MAYBRJC02403.1
(1,1,2,2-Tetrafluorethoxy)benzol
(1,1,2,2-Tetrafluorethoxy)benzol
Formel: HF₂CCF₂OC₆H₅
Molecular Weight: 194,13 g/mol
MDL: MFCD00042102
CAS-Nummer: 350-57-2
EINECS: 206-505-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca7b4a8c5103a98058318de8cc3178a31c7e3ebe&vid=90f4f916b50991991e83faa034e58e98db533476&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR