5-Brom-2-iodtoluol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-4-iodo-3-methylbenzene

HC-3025-5G HC-3025-25G HC-3025-100G
COBBHC-3025-5GEA 25.5 EUR
COBBHC-3025-5G COBBHC-3025-25G COBBHC-3025-100G
5-Brom-2-iodtoluol
5-Brom-2-iodtoluol
Formel: C₇H₆BrI
Molecular Weight: 296,93 g/mol
MDL: MFCD00060664
CAS-Nummer: 116632-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR