4-Aminobenzamidoxim

Lieferant: Cayman Chemical

Synonyme: 4-Aminobenzamide oxime, 4-Amino-N'-hydroxybenzimidamide

11053-500 11053-5 11053-1
CAYM11053-500EA 44 EUR
CAYM11053-500 CAYM11053-5 CAYM11053-1
4-Aminobenzamidoxim
4-Aminobenzamidoxime
Formel: C₇H₉N₃O
Molecular Weight: 151,17 g/mol
MDL: MFCD08061151
CAS-Nummer: 277319-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR