3,3-Dimethylglutarimid

Lieferant: CHEMPUR

Synonyme: 4,4-dimethylpiperidine-2,6-dione

XBX00216.250 XBX00216.1
CHMPXBX00216.250EA 0 EUR
CHMPXBX00216.250 MAYBXBX00216.1
3,3-Dimethylglutarimid
3,3-Dimethylglutarimid
Formel: C₇H₁₁NO₂
Molecular Weight: 141,17 g/mol
MDL: MFCD00006671
CAS-Nummer: 1123-40-6
EINECS: 214-373-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR