2-(Trimethylacetyl)thiophen ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2-Dimethyl-1-(thiophen-2-yl)propan-1-one
L07560.06
L07560.14
L07560.06EA
87.4
EUR
L07560.06
L07560.14
2-(Trimethylacetyl)thiophen ≥98%
2-(Trimethylacetyl)thiophen
Purity: 98%
CAS No.: 20409-48-7
Molecular Formula: C9H12OS
Formula Weight: 168.26
Flash Point: 99°(210°F)
Refractive Index: 1.5350
Boiling Point: 114-116°/14mm
Beilstein Reference: 116243
MDL No.: MFCD00051668
Formel:
C₉H₁₂OS Molecular Weight: 168,26 g/mol Siedepunkt: 114…116 °C (14 mmHg) Dichte: 1,07 Flammpunkt: 99 °C (210 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051668 CAS-Nummer: 20409-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...