Methylphenylglyoxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl benzoylformate, Methyl 2-oxo-2-phenylacetate

M-Clarity™ Qualitätsstufe = MQ100

M30507-25G M30507-5G
SIALM30507-25GEA 123 EUR
SIALM30507-25G SIALM30507-5G
Methylphenylglyoxylat, Sigma-Aldrich®
Methylphenylglyoxylat
Formel: C₉H₈O₃
Molecular Weight: 164,1600 g/mol
MDL: MFCD00008443
CAS-Nummer: 15206-55-0
EINECS: 239-263-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR