cis,cis,cis-13,16,19-Docosatriensäure

Lieferant: Cayman Chemical

90170-100 90170-50 90170-25
CAYM90170-100EA 171 EUR
CAYM90170-100 CAYM90170-50 CAYM90170-25
cis,cis,cis-13,16,19-Docosatriensäure
cis,cis,cis-13,16,19-Docosatrienoic acid
Formel: C₂₂H₃₈O₂
CAS-Nummer: 28845-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR