3-Brom-5-fluor-4-methoxyanilin
Lieferant: COMBI-BLOCKS
AN-1232-5G
AN-1232-1G
COBBAN-1232-5GEA
580
EUR
COBBAN-1232-5G
COBBAN-1232-1G
3-Brom-5-fluor-4-methoxyanilin
3-Brom-5-fluor-4-methoxyanilin
Formel:
C₇H₇BrFNO Molecular Weight: 220,04 g/mol |
MDL:
MFCD08062384 CAS-Nummer: 875664-44-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...