Bis(cyclopentadienyl)cobalt, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cobaltocene, Bis(cyclopentadienyl)cobalt(II)
M-Clarity™ Qualitätsstufe = MQ100
339164-10G
339164-2G
SIAL339164-10GEA
695
EUR
SIAL339164-10G
SIAL339164-2G
Bis(cyclopentadienyl)cobalt, Sigma-Aldrich®
Bis(cyclopentadienyl)cobalt
Formel:
C₁₀H₁₀Co Molecular Weight: 189.12 g/mol |
MDL:
MFCD00013749 CAS-Nummer: 1277-43-6 EINECS: 215-061-0 UN: 3181 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...