2-Brom-1-chlor-4-iodbenzol

Lieferant: COMBI-BLOCKS

HC-3042-5G HC-3042-1G
COBBHC-3042-5GEA 208 EUR
COBBHC-3042-5G COBBHC-3042-1G
2-Brom-1-chlor-4-iodbenzol
2-Brom-1-chlor-4-iodbenzol
Formel: C₆H₃BrCl
Molecular Weight: 317,3495 g/mol
MDL: MFCD07780667
CAS-Nummer: 31928-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR