3-(2-Bromacetyl)thiophen

Lieferant: BIONET RESEARCH

10X-0708.1 10X-0708.5 10X-0708.500
BIOT10X-0708.1EA 0 EUR
BIOT10X-0708.1 BIOT10X-0708.5 BIOT10X-0708.500
3-(2-Bromacetyl)thiophen
3-(2-Bromacetyl)thiophen
Formel: C₆H₅BrOS
CAS-Nummer: 1468-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR