2-Acetyl-3,4-dibromthiophen
Lieferant: BIONET RESEARCH
Synonyme:
1-(3,4-Dibromothiophen-2-yl)ethanone, 1-(3,4-Dibromothien-2-yl)ethanone
9Y-0810.1
9Y-0810.5
9Y-0810.500
BIOT9Y-0810.1EA
0
EUR
BIOT9Y-0810.1
BIOT9Y-0810.5
BIOT9Y-0810.500
2-Acetyl-3,4-dibromthiophen
2-Acetyl-3,4-dibromthiophen
Formel:
C₆H₄Br₂OS |
CAS-Nummer:
57681-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...