3-Brom-2-(2-bromacetyl)thiophen

Lieferant: BIONET RESEARCH

Synonyme: 2-Bromo-1-(3-bromothiophen-2-yl)ethan-1-one

9J-901.500 9J-901.1 9J-901.5 9J-901.10
BIOT9J-901.500EA 0 EUR
BIOT9J-901.500 BIOT9J-901.1 BIOT9J-901.5 BIOT9J-901.10
3-Brom-2-(2-bromacetyl)thiophen
3-Brom-2-(2-bromacetyl)thiophen
Formel: C₆H₄Br₂OS
CAS-Nummer: 81216-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR