3-(4-Hydroxyphenyl)propiononitril

Lieferant: CHEMPUR

Synonyme: 3-(4-Hydroxyphenyl)propionitrile

RJC02789.250 RJC02789.1
CHMPRJC02789.250EA 0 EUR
CHMPRJC02789.250 MAYBRJC02789.1
3-(4-Hydroxyphenyl)propiononitril
3-(4-Hydroxyphenyl)propiononitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00016492
CAS-Nummer: 17362-17-3
EINECS: 241-393-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR