Methyl-α-D-glucopyranosid 99%

Lieferant: Thermo Scientific

Synonyme: α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

173841000 173845000 173840025
ACRO173841000EA 32.1 EUR
ACRO173841000 ACRO173845000 ACRO173840025
Methyl-α-D-glucopyranosid 99%
Methyl-α-D-glucopyranosid
Formel: C₇H₁₄O₆
Molecular Weight: 194,18 g/mol
Siedepunkt: 200 °C (,2 mmHg)
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064086
CAS-Nummer: 97-30-3
EINECS: 202-571-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.1 % (60 to 70°C)
HPLC ≥98.5 % (ELSD)
Specific optical rotation +156° to +161° (20°C, 589 nm) (c=10, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR