Methyl-α-D-glucopyranosid ≥98%
Lieferant: Thermo Scientific
Synonyme:
α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
A12484.22
A12484.36
A12484.22EA
32
EUR
A12484.22
A12484.36
Methyl-α-D-glucopyranosid ≥98%
Methyl-α-D-glucopyranosid
Purity: 98+%
CAS No.: 97-30-3
Molecular Formula: C7H14O6
Formula Weight: 194.18
Melting Point: 166-171°
Solubility: Soluble in water. Insoluble in ether
Beilstein Reference: 81568
Merck Reference: 14,6080
Optical Rotation: +159° (c=10 in water)
MDL No.: MFCD00064086
Formel:
C₇H₁₄O₆ Molecular Weight: 194,18 g/mol Schmelzpunkt: 166…171 °C Dichte: 1,460 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064086 CAS-Nummer: 97-30-3 EINECS: 202-571-3 Merck Index: 14,6080 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...