(4-Nitrophenyl)-α-D-glucopyranosid, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Synonyme:
4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside
487506-1
487506-5
487506-1EA
56
EUR
487506-1
487506-5
(4-Nitrophenyl)-α-D-glucopyranosid, Sigma-Aldrich®
(4-Nitrophenyl)-α-D-glucopyranosid
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301,25 g/mol |
MDL:
MFCD00064088 CAS-Nummer: 3767-28-0 EINECS: 223-189-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...