4-(Trifluormethoxy)anilin Hydrochlorid

Lieferant: CHEMPUR

RF05447.250 RF05447DA
CHMPRF05447.250EA 0 EUR
CHMPRF05447.250 MAYBRF05447DA
4-(Trifluormethoxy)anilin Hydrochlorid
4-(Trifluormethoxy)anilin Hydrochlorid
Formel: C₇H₇ClF₃NO
MDL: MFCD00219956
CAS-Nummer: 42823-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR