7-Chloroxindol
Lieferant: COMBI-BLOCKS
Synonyme:
7-Chloroindolin-2-one
IN-0110-5G
IN-0110-25G
IN-0110-1G
COBBIN-0110-5GEA
302
EUR
COBBIN-0110-5G
COBBIN-0110-25G
COBBIN-0110-1G
7-Chloroxindol
7-Chloroxindol
Formel:
C₈H₆ClNO Molecular Weight: 167,5923 g/mol |
MDL:
MFCD06656001 CAS-Nummer: 25369-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...