7-Fluoroxindol

Lieferant: COMBI-BLOCKS

Synonyme: 7-Fluoroindolin-2-one

IN-0087-1G IN-0087-5G
COBBIN-0087-1GEA 90.9 EUR
COBBIN-0087-1G COBBIN-0087-5G
7-Fluoroxindol
7-Fluoroxindol
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD02179608
CAS-Nummer: 71294-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR