1-Methyloxindol

Lieferant: Thermo Scientific

MAYBRH01320.1EA 0 EUR
MAYBRH01320.1 CHMPRH01320.250
1-Methyloxindol
1-Methyloxindol
Formel: C₉H₉NO
Molecular Weight: 147,1739 g/mol
MDL: MFCD00030253
CAS-Nummer: 61-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR