1,3-Bis(1,1,2,2-tetrafluorethoxy)benzol

Lieferant: FLUOROCHEM

010420-10G 010420-25G
FLUO010420-10GEA 0 EUR
FLUO010420-10G FLUO010420-25G
1,3-Bis(1,1,2,2-tetrafluorethoxy)benzol
1,3-Bis(1,1,2,2-tetrafluorethoxy)benzol
Formel: C₁₀H₆F₈O₂
MDL: MFCD00497882
CAS-Nummer: 3914-19-0
EINECS: 223-476-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR