N-Methylhydroxylammoniumchlorid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Methylhydroxylamine hydrochloride
L02202.06
L02202.14
L02202.06EA
46.5
EUR
L02202.06
L02202.14
N-Methylhydroxylammoniumchlorid 98%
N-Methylhydroxylammoniumchlorid
Purity: 98%
CAS No.: 4229-44-1
Molecular Formula: CH3NHOH.HCl
Formula Weight: 83.52
Melting Point: 80-86°
Beilstein Reference: 3541409
Formel:
CH₆ClNO Molecular Weight: 83.52 g/mol Schmelzpunkt: 80…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012597 CAS-Nummer: 4229-44-1 EINECS: 224-181-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...