N-Methylhydroxylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Methylhydroxylamine hydrochloride
M-Clarity™ Qualitätsstufe = MQ200
M50400-10G
M50400-50G
M50400-5G
SIALM50400-10GEA
99.8
EUR
SIALM50400-10G
SIALM50400-50G
SIALM50400-5G
N-Methylhydroxylammoniumchlorid, Sigma-Aldrich®
N-Methylhydroxylammoniumchlorid
Formel:
CH₆ClNO Molecular Weight: 83,52 g/mol |
MDL:
MFCD00012597 CAS-Nummer: 4229-44-1 EINECS: 224-181-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...