2-Ethyl-6-methylphenol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
B25213.03
B25213.06
B25213.14
B25213.03EA
33.3
EUR
B25213.03
B25213.06
B25213.14
2-Ethyl-6-methylphenol ≥97%
2-Ethyl-6-methylphenol
Purity: 97%
CAS No.: 1687-64-5
Molecular Formula: C9H12O
Formula Weight: 136.19
Refractive Index: 1.5310
Formel:
C₉H₁₂O Molecular Weight: 136,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00962292 CAS-Nummer: 1687-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...