2,4-Dimethyl-6-(2-methyl-2-propanyl)phenol

Lieferant: Thermo Scientific

Synonyme: 2-(tert-Butyl)-4,6-dimethylphenol

SB01223EA SB01223EE
MAYBSB01223EAEA 295 EUR
MAYBSB01223EA MAYBSB01223EE
2,4-Dimethyl-6-(2-methyl-2-propanyl)phenol
6-tert-Butyl-2,4-xylenol
Formel: C₁₂H₁₈O
Molecular Weight: 178,28 g/mol
MDL: MFCD00002234
CAS-Nummer: 1879-09-0
EINECS: 217-533-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR