(1-Phenylethyliden)malononitril

Lieferant: BIONET RESEARCH

Synonyme: 2-(1-Phenylethylidene)propanedinitrile, 2-(1-Phenylethylidene)malononitrile

2R-0229.500 2R-0229.1 2R-0229.5 2R-0229.10
BIOT2R-0229.500EA 0 EUR
BIOT2R-0229.500 BIOT2R-0229.1 BIOT2R-0229.5 BIOT2R-0229.10
(1-Phenylethyliden)malononitril
(1-Phenylethyliden)malononitril
Formel: C₁₁H₈N₂
Molecular Weight: 168,1946 g/mol
MDL: MFCD00159012
CAS-Nummer: 5447-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR