Hydroxylammoniumchlorid 7% in wässriger Lösung, reinst

Lieferant: Bernd Kraft

Synonyme: Hydroxylamin HydrochloridChlorhydrathydroxylaminHydroxylamin HClHydroxylamin-Hydrochlorid

KRAF01720.2000EA 59.7 EUR
KRAF01720.2000 KRAF01720.3000 KRAF01720.5000
Hydroxylammoniumchlorid 7% in wässriger Lösung, reinst
Hydroxylammoniumchlorid
Formel: NH₂OH·HCl
Molecular Weight: 69,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051089
CAS-Nummer: 5470-11-1
EINECS: 226-798-2
UN: 1789
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR