N,O-Dimethylhydroxylamin Hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
Achtung
A17469.06 A17469.14 A17469.22
A17469.06EA 34.3 EUR
A17469.06 A17469.14 A17469.22
N,O-Dimethylhydroxylamin Hydrochlorid 98%
N,O-Dimethylhydroxylamin Hydrochlorid
Purity: 98% CAS No.: 6638-79-5 Molecular Formula: CH3ONHCH3.HCl Formula Weight: 97.55 Melting Point: 112-115° Beilstein Reference: 3650353
Formel: C₂H₈ClNO
Molecular Weight: 97.55 g/mol
Schmelzpunkt: 112…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012485
CAS-Nummer: 6638-79-5
EINECS: 229-642-1