N,O-Dimethylhydroxylamin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
D163708-100G
D163708-25G
D163708-5G
SIALD163708-100GEA
272
EUR
SIALD163708-100G
SIALD163708-25G
SIALD163708-5G
N,O-Dimethylhydroxylamin Hydrochlorid, Sigma-Aldrich®
N,O-Dimethylhydroxylamin Hydrochlorid
Formel:
C₂H₈ClNO Molecular Weight: 97.55 g/mol |
MDL:
MFCD00012485 CAS-Nummer: 6638-79-5 EINECS: 229-642-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...