Oxindol-6-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one
PN-5414-1G
PN-5414-250M
COBBPN-5414-1GEA
385
EUR
COBBPN-5414-1G
COBBPN-5414-250M
Oxindol-6-boronsäurepinakolester
Oxindol-6-boronsäurepinakolester
Formel:
C₁₄H₁₈BNO₃ Molecular Weight: 259,1086 g/mol |
MDL:
MFCD11855965 CAS-Nummer: 893441-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...