Methyloxindol-5-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl Oxindole-5-carboxylate

IN-0074-5G IN-0074-1G
COBBIN-0074-5GEA 230 EUR
COBBIN-0074-5G COBBIN-0074-1G
Methyloxindol-5-carboxylat
Methyloxindol-5-carboxylat
Formel: C₁₀H₉NO₃
Molecular Weight: 191,1834 g/mol
MDL: MFCD02179600
CAS-Nummer: 199328-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR