Oxindol-5-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Oxoindoline-5-carboxylic Acid, 2-Oxindole-5-carboxylic acid
IN-0070-5G
IN-0070-1G
COBBIN-0070-5GEA
358
EUR
COBBIN-0070-5G
COBBIN-0070-1G
Oxindol-5-carbonsäure
Oxindol-5-carbonsäure
Formel:
C₉H₇NO₃ Molecular Weight: 177,1600 g/mol |
MDL:
MFCD03411961 CAS-Nummer: 102359-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...