6-(Methylsulfonyl)oxindol

Lieferant: COMBI-BLOCKS

IN-0084-5G IN-0084-1G
COBBIN-0084-5GEA 1020 EUR
COBBIN-0084-5G COBBIN-0084-1G
6-(Methylsulfonyl)oxindol
6-(Methylsulfonyl)oxindol
Formel: C₉H₉NO₃S
MDL: MFCD02179607
CAS-Nummer: 850429-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR