Tetrabutylammoniumdihydrogenphosphat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

M-Clarity™ Qualitätsstufe = MQ100

86833-25G 86833-5G
SIAL86833-25GEA 737 EUR
SIAL86833-25G SIAL86833-5G
Tetrabutylammoniumdihydrogenphosphat, Sigma-Aldrich®
Tetrabutylammoniumdihydrogenphosphat
Formel: C₁₆H₃₈NO₄P
MDL: MFCD00064526
CAS-Nummer: 5574-97-0
EINECS: 226-947-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR