2-Brompyrazin

Lieferant: COMBI-BLOCKS

PY-7784-1G PY-7784-5G
COBBPY-7784-1GEA 88.3 EUR
COBBPY-7784-1G COBBPY-7784-5G
2-Brompyrazin
2-Bromopyrazine
Formel: C₄H₃BrN₂
Molecular Weight: 158.9800 g/mol
MDL: MFCD08275680
CAS-Nummer: 56423-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR