2-Amino-5-brompyrazin

Lieferant: Thermo Scientific

Synonyme: 5-bromopyrazin-2-amine

MO00943DA MO00943-0001
MAYBMO00943DAEA 0 EUR
MAYBMO00943DA ABCRMO00943-0001
2-Amino-5-brompyrazin
2-Amino-5-bromopyrazine
Formel: C₄H₄BrN₃
Molecular Weight: 174.00 g/mol
MDL: MFCD00235015
CAS-Nummer: 59489-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR