2-Amino-5-brom-3-(methylamino)pyrazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

696110-1G 696110-250MG
SIAL696110-1GEA 356 EUR
SIAL696110-1G SIAL696110-250MG
2-Amino-5-brom-3-(methylamino)pyrazin, Sigma-Aldrich®
2-Amino-5-bromo-3-(methylamino)pyrazine
Formel: C₅H₇BrN₄
CAS-Nummer: 55635-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR